Learn More
Strem, An Ascensus Company CAS# 854045-93-5. 1g. Chloro[2-(di-t-butylphosphino))-1,1'-biphenyl]gold(I), 99%. MFCD09265131
SDP
Supplier:  Strem, An Ascensus Company 7903401G
Encompass
CAS# 854045-93-5. 1g. Chloro[2-(di-t-butylphosphino))-1,1'-biphenyl]gold(I), 99%. MFCD09265131. Molecular Weight: 530.82. Molecular Formula: C20H27AuClP. Color/form: white pwdr. Strem# 79-0340. http://www.strem.com/catalog/v/79-0340/
Catalog No. 50-901-13386