Learn More
Strem, An Ascensus Company CAS# 32993-05-8. 1g. Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II), 99%. MFCD00075004
SDP
Supplier:  Strem, An Ascensus Company 4401201G
Encompass
CAS# 32993-05-8. 1g. Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II), 99%. MFCD00075004. Molecular Weight: 726.20. Molecular Formula: C5H5Ru[P(C6H5)3]2Cl. Color/form: orange xtl. Strem# 44-0120. http://www.strem.com/catalog/v/44-0120/
Catalog No. 50-901-11797