Learn More
Medchemexpress LLC Chloro-L-alanine | 2731-73-9 | MFCD00046536 | 98.0% | 123.54 g/mol | C3H6ClNO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10737310MM 1ML
Encompass_Preferred
β-Chloro-L-alanine is a halogenated amino acid analog used in biochemical and microbiology research. It inhibits enzymes such as threonine deaminase and alanine racemase and is supplied as a 10 mM aqueous solution (1 mL) for laboratory assays. The compound has CAS 2731-73-9, molecular formula C3H6ClNO2, molecular weight 123.54 g/mol, and typical purity 98.0%.
  • Bacteriostatic amino acid analog useful for enzyme inhibition studies.
  • Inhibits threonine deaminase and alanine racemase.
  • Provided as a 10 mM aqueous solution, 1 mL volume.
  • High purity (98.0%) suitable for research applications.
  • Water soluble for easy preparation in assays.
Catalog No. 50-002-40862