Learn More
Strem, An Ascensus Company CAS# 944451-28-9. 1g. Chloro[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-segphos)]Cl. MFCD09753017
SDP
Supplier:  Strem, An Ascensus Company 4400961G
Encompass
CAS# 944451-28-9. 1g. Chloro[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-segphos)]Cl. MFCD09753017. Molecular Weight: 916.77. Molecular Formula: [C48H42ClO4P2Ru]+Clae. Color/form: yellow pwdr. Strem# 44-0096. http://www.strem.com/catalog/v/44-0096/
Catalog No. NC1773780