Learn More
Medchemexpress LLC Chloroacetamido-PEG4-C2-Boc | 1365655-90-8 | ≥97.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130470100MG
Encompass_Preferred
Chloroacetamido-PEG4-C2-Boc is a PROTAC linker, a molecule designed to induce the degradation of target proteins. It belongs to the PEG and Alkyl/ether class, and its tetraethylene glycol component can be utilized in the synthesis of various PROTAC molecules.
  • Acts as a PEG/Alkyl/ether-based PROTAC linker
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase, the other for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40817