Learn More
Medchemexpress LLC Chloroacetamido-PEG4-C2-Boc | 1365655-90-8 | ≥97.0% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY130470
Encompass_Preferred
Chloroacetamido-PEG4-C2-Boc is a PROTAC linker that belongs to the PEG and Alkyl/ether class. This linker, which includes tetraethylene glycol, is suitable for the synthesis of various PROTAC molecules.
  • Used in the synthesis of PROTACs
  • PROTACs connect two distinct ligands: one targeting an E3 ubiquitin ligase, the other a specific protein
  • Leverages the intracellular ubiquitin-proteasome system
  • Facilitates the selective degradation of target proteins
Catalog No. 50-003-41495