Learn More
Abcam Chloramphenicol ß-D-galactoside, 2MG
Supplier:  Abcam AB2750712MG
Encompass_Preferred
MW 485.3 Da. Chloramphenicol precursor that releases chloramphenicol upon enzymatic or chemical hydrolysis. Galactose is linked to the site of action of chloramphenicol via a hydrolysable bond.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-075