Learn More
Medchemexpress LLC Chlorobutanol (hemihydrate) | 6001-64-5 | MFCD02179352 | 100.0% | 186.46 g/mol | C4H7Cl3O · 1/2 H2O | 10 G

Supplier: Medchemexpress LLC HYW08985610G
Chlorobutanol (hemihydrate) is an antimicrobial preservative and analytical standard supplied in the hemihydrate form. It is provided with batch-specific purity documentation and a certificate of analysis for laboratory and pharmaceutical applications.
- Form: hemihydrate crystalline solid
- Purity: batch-specific assay with COA
- Intended use: preservative and analytical reference standard
- CAS number: 6001-64-5
- Molecular weight: 186.46 g/mol
- Pack size: 10 G
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.