Learn More
Medchemexpress LLC Chlorocholine chloride | 999-81-5 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB188610MM/1ML
Encompass_Preferred
Chlorocholine chloride is an effective plant growth regulator that inhibits gibberellin (GA) biosynthesis. It is suitable for research into the mechanisms and effects of GA on plant cell growth and differentiation.
  • Inhibits gibberellin (GA) biosynthesis
  • Increases the biomass of leaves and stems
  • Enhances sucrose content in leaves
  • Decreases gibberellic acid (GA) and increases indole-3-acetic acid (IAA) contents in lily bulbs
Catalog No. 50-004-10823