Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroform suitable for HPLC, >=99.8%, amylene stabilized | 67-66-3 | MFCD00000826 | 1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 348541L
Encompass
Chloroform suitable for HPLC, >=99.8%, amylene stabilized | Purity: >=99.8% | Mol Wt: 119.38 | 67-66-3 | MFCD00000826 | 1L
Catalog No. NC1803034