Learn More
Apexbio Technology LLC Chlorogenic acid 327-97-9 500mg
SDP
Supplier:  Apexbio Technology LLC N1769500
Encompass_Preferred
Chlorogenic acid (327-97-9) is a small-molecule modulator primarily involved in the regulation of oxidative stress and inflammation It acts by scavenging reactive oxygen species (ROS) and inhibiting inflammatory pathways including suppression of cytokine expression and NF- B signaling Chlorogenic acid also exhibits antiviral activity through inhibition of viral entry and replication as demonstrated in cell-culture assays Reported IC50 values typically range from approximately 40 80 M depending on experimental conditions cell types and bioassays Based on these pharmacological properties chlorogenic acid holds research potential in oxidative stress-related disorders inflammation pathogenesis viral pathogenicity and hepatotoxicity models
Catalog No. 50-224-0306