Learn More
Medchemexpress LLC Chloromethyl methyl carbonate | 40510-81-4 | 99.3% | 124.52 | 10 G
SDP
Supplier:  Medchemexpress LLC HY4338410G
Encompass_Preferred
Chloromethyl methyl carbonate is a chemical reagent primarily used as a laboratory chemical and in the manufacture of substances. This product is classified as a highly flammable liquid and vapor, harmful if swallowed, and can cause severe skin burns and eye damage. It is intended for research use only and must be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities.
  • Used as a laboratory chemical and for the manufacture of substances
  • Highly flammable liquid and vapor
  • Harmful if swallowed
  • Can cause severe skin burns and eye damage
  • Stable under recommended storage conditions of 4°C, sealed, away from moisture and light
  • Liquid appearance
  • Boiling point/range of 139-140 °C at 760 mmHg
  • Relative density of 1.303 g/cm³
Catalog No. 50-004-04701