Learn More
Medchemexpress LLC Chloroneb 25g | 2675-77-6 | 207.06 g/mol | C8H8Cl2O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11699225G
Encompass_Preferred
Chloroneb is an analytical reference standard (Demosan) used for pesticide residue analysis and environmental testing. It is a chlorinated dimethoxybenzene with formula C8H8Cl2O2 and a molecular weight around 207 g/mol.
  • Analytical reference standard for pesticide residue detection.
  • Suitable for GC and qNMR assay-based analysis.
  • Available in small pack sizes for analytical workflows.
  • Storage: stable at room temperature; in solvent, store at -80°C for 2 years or -20°C for 1 year.
  • For research use only; not validated for medical applications.
Catalog No. 50-002-27290