Learn More
Medchemexpress LLC Chloroneb 50g | 2675-77-6 | 207.05 g/mol | C8H8Cl2O2 | 50 G

Supplier: Medchemexpress LLC HY11699250G
Chloroneb (Demosan) is a chlorinated dimethoxybenzene fungicidal pesticide supplied as an analytical standard for residue analysis in food and environmental samples. It is used to detect organochlorine pesticide residues with good selectivity, stability, and reproducibility in validated analytical methods.
- Supplied as an analytical standard for residue detection
- Purity ≥95.0%
- Suitable for analysis of water, soil, and food samples
- Available in a 50 g pack for bulk analytical use
- Molecular formula C8H8Cl2O2; molecular weight ≈207.05 g/mol
- Stable and reproducible in extraction and detection workflows
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.