Learn More
Medchemexpress LLC Chlorophosphonazo III | 1914-99-4 | MFCD00042698 | 757.36 g/mol | C22H16Cl2N4O14P2S2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW250149500MG
Encompass_Preferred
Chlorophosphonazo III is a colorimetric reagent used for spectrophotometric determination of calcium and magnesium and for detection of various metal ions. Supplied as a solid powder, it is used in biochemical assays to measure alkaline earth metals and certain transition and actinide elements under pH-specific conditions.
  • Colorimetric reagent for calcium and magnesium determination
  • Effective at pH-specific measurements (calcium at pH 2.2; calcium and magnesium at pH 7.0)
  • Suitable for spectrophotometric detection of titanium, zirconium, thorium, scandium, and select actinides
  • Supplied as a solid powder in laboratory pack sizes, including 500 mg
  • Accompanied by product datasheet and safety data sheet for handling guidance
Catalog No. 50-002-80873