Learn More
Medchemexpress LLC Chlorophosphonazo III | 1914-99-4 | MFCD00042698 | 93.1% | 757.36 g·mol⁻¹ | C22H16Cl2N4O14P2S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW250149100MG
Encompass_Preferred
Chlorophosphonazo III is a colorimetric reagent for spectrophotometric determination of alkaline earth metals and other metal ions. It measures calcium at pH 2.2 and both calcium and magnesium at pH 7.0, and is used for laboratory assays and analytical determinations. Intended for research use only.
  • Sensitive reagent for calcium and magnesium detection.
  • Allows calcium-only measurement at pH 2.2.
  • Allows simultaneous calcium and magnesium measurement at pH 7.0.
  • Applicable to spectrophotometric determination of titanium, zirconium, thorium, scandium, and certain actinides.
  • Provided in laboratory pack sizes including 100 mg.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-80711