Learn More
Apexbio Technology LLC Chloroquine 54-05-7 200mg
SDP
Supplier:  Apexbio Technology LLC BA1002200
Encompass_Preferred
Chloroquine (CAS 54-05-7) is a small molecule known for its anti-inflammatory properties and is widely used in biomedical research It functions primarily by inhibiting autophagy through disruption of lysosomal acidification and by modulating Toll-like receptor signaling pathways Chloroquine has demonstrated potent inhibitory effects on infection with reported activity at concentrations as low as 1 13 M Its mechanisms make it a valuable tool for investigating processes related to inflammation immune modulation and host pathogen interactions in cellular and molecular studies
Catalog No. 50-000-89337