Learn More
Medchemexpress LLC Chloroquine phosphate | 50-63-5 | MFCD00025732 | 99.89% | 515.86 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY17589200MG
Encompass_Preferred
Chloroquine phosphate is an antimalarial and anti-inflammatory agent widely used to treat malaria and rheumatoid arthritis. Chloroquine phosphate is an autophagy and toll-like receptors (TLRs) inhibitor. Chloroquine phosphate is highly effective in the control of SARS-CoV-2 (COVID-19) infection in vitro (EC50=1.13 μM).
  • Antimalarial and anti-inflammatory agent
  • Used to treat malaria and rheumatoid arthritis
  • Autophagy and toll-like receptors (TLRs) inhibitor
  • Highly effective in the control of SARS-CoV-2 (COVID-19) infection in vitro (EC50=1.13 μM)
Catalog No. 50-003-87529