Learn More
Selleck Chemical LLC Chloroquine100mg
Supplier: Selleck Chemical LLC S6999100MG
Chloroquine (NSC-187208, Aralen, CHQ, CQ) is an antimalarial drug andautophagy/lysosome inhibitor. Chloroquine also suppressesToll-like receptor-9 (TLR9) protein expression. Chloroquine is highly effective agianst SARS-CoV-2 (COVID-19) infection with EC50 of 1.13 ?M in Vero E6 cells. Chloroquine has anti-HIV-1 activity. *For Research & Development use only. Product is not intended for drug, household, or human consumption.