Learn More
Medchemexpress LLC Chloroquinoxaline sulfonamide | 97919-22-7 | 99.1% | 334.78 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY106662
Encompass_Preferred
Chloroquinoxaline sulfonamide is a structural analogue of sulfaquinoxaline and functions as a topoisomerase II alpha/beta poison. It is used to control coccidiosis in poultry, rabbit, sheep, and cattle, and also exhibits antitumor activity.
  • Structural analogue of sulfaquinoxaline
  • Topoisomerase II alpha/beta poison
  • Used to control coccidiosis in poultry, rabbit, sheep, and cattle
  • Exhibits antitumor activity
  • Causes dose-dependent protein-DNA cross-links
  • Inhibits proliferation of murine B16 melanoma cells (at 1 mM concentration)
Catalog No. 50-003-66002