Learn More
Cayman Chemical ChlorothIazIde 10g
SDP
Supplier:  Cayman Chemical 1790910 G
Encompass_Preferred
A thiazide diuretic that increases renal excretion of sodium, potassium, chloride, and bicarbonate ions by inhibiting tubular reabsorptive mechanisms
Catalog No. 50-296-5726