Learn More
Medchemexpress LLC Chlorotrianisene | 569-57-3 | 99.02% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB215810MM/1ML
Encompass_Preferred
Chlorotrianisene is a long-acting non-steroidal estrogen and an orally active estrogen receptor modulator. It exhibits antiestrogenic activity and potently inhibits the enzyme COX-1, also inhibiting platelet aggregation in whole blood.
  • Long-acting non-steroidal estrogen
  • Orally active estrogen receptor modulator
  • Exhibits antiestrogenic activity
  • Potently inhibits the enzyme COX-1
  • Inhibits platelet aggregation in whole blood
  • Available as a 10 mM solution in DMSO
  • Purity of 99.02%
Catalog No. 50-004-10469