Learn More
Medchemexpress LLC Chloroxuron-d6 | 2030182-44-4 | 98.75% | C15H9D6ClN2O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY143936S1MG
Encompass_Preferred
Chloroxuron-d6 is the deuterium labeled Chloroxuron. It can be used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Chloroxuron
  • Used as a tracer for quantitative analysis
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-56159