Learn More
U.S. Pharmacopeia Chloroxylenol, 88-04-0, MFCD00002324, 125mg
Supplier:  U.S. Pharmacopeia 1122700
Encompass
Molecular formula C8H9ClO, Molecular weight 156.61, Melting Point 239.9 °F (115.5 °C), Boiling Point 474.8 °F (246 °C), Synonyms: Parachlorometaxylenol, PCMX
Catalog No. NC0061331