Learn More
Cayman Chemical ChlorphenIramIn-d6maleate 5mg
SDP
Supplier:  Cayman Chemical 357675 MG
Encompass_Preferred
An internal standard for the quantification of chlorpheniramine by GC- or LC-MS
Catalog No. 50-291-0571