Learn More
Medchemexpress LLC Chlorpheniramine maleate | 113-92-8 | HY-B0286A | 99.7% | 390.86 | 1 G
SDP
Supplier:  Medchemexpress LLC 1GHYB0286A
Encompass_Preferred
Chlorpheniramine maleate is a histamine H1 receptor antagonist. It is a well-characterized compound that shows antimalarial activity against Plasmodium falciparum and inhibits scratching in Ovalbumin-challenged BALB/c mice. This product is suitable for a wide range of research applications in inflammation, immunology, and neuronal signaling.
  • Histamine H1 receptor antagonist
  • Antimalarial activity
  • Inhibits scratching in mice
  • Soluble in DMSO, H2O, and PBS
  • High purity of 99.7% (LCMS)
Catalog No. 50-003-65399