Learn More
Medchemexpress LLC Chlorpromazine-d6 hydrochloride | 1228182-46-4 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0407AS10MM/1ML
Encompass_Preferred
Chlorpromazine-d6 (hydrochloride) is the deuterium labeled Chlorpromazine. Chlorpromazine is an inhibitor of dopamine receptor, 5-HT receptor, potassium channel, and sodium channel.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09172