Learn More
Selleck Chemical LLC Chlorpromazine HCl 50mg 69-09-0 Sonazine
SDP
Supplier:  Selleck Chemical LLC S245650MG
Encompass_Preferred
Chlorpromazine HCl (Sonazine) is a dopamine and potassium channel inhibitor with IC50 of 6.1 and 16 ?M for inward-rectifying K+ currents and time-independent outward currents. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2422