Learn More
Selleck Chemical LLC Chlorpromazine HCl S2456-1g
SDP
Supplier:  Selleck Chemical LLC S24561G
Encompass_Preferred
Chlorpromazine HCl is a dopamine and potassium channel inhibitor with IC50 of 6 1 and 16 M for inward-rectifying K currents and time-independent outward currents
Catalog No. 50-311-2695