Learn More
Medchemexpress LLC Chlorpropham | 101-21-3 | MFCD00037108 | 95.9% | 213.66 g·mol⁻¹ | C10H12ClNO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01424010G
Encompass_Preferred
Chlorpropham is a carbamate herbicide and plant growth regulator used in research and analytical applications. It inhibits mitosis and cell division by interfering with the organisation of spindle microtubules and is supplied for laboratory use in both solid and solution forms.
  • Carbamate herbicide and plant growth regulator.
  • Inhibits mitosis by disrupting spindle microtubule organization.
  • Provided as solid powder and as DMSO solutions for ready use.
  • Suitable as an analytical standard for research applications.
  • Typical purity around 95.9% for the solid material.
Catalog No. 50-003-04010