Learn More
Medchemexpress LLC Chlorprothixene (hydrochloride) | 6469-93-8 | 99.9% | 200 MG

Supplier: Medchemexpress LLC HYB0274A200MG
Chlorprothixene hydrochloride is an antagonist of dopamine and histamine receptors, demonstrating antipsychotic activity. It exhibits Ki values of 18 nM for hD1, 2.96 nM for hD2, 4.56 nM for hD3, 9 nM for hD5, and 3.75 nM for hH1 receptors.
- Antagonist for dopamine and histamine receptors
- Exhibits antipsychotic activity
- Binds to 5-HT receptors
- Molecular weight: 352.32
- Chemical formula: C18H19Cl2NS
- Appearance: White to off-white solid
- Purity: 99.91%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.