Learn More
Medchemexpress LLC Chlorprothixene (hydrochloride) | 6469-93-8 | 99.9% | 50 MG

Supplier: Medchemexpress LLC HYB0274A50MG
Chlorprothixene hydrochloride is an antagonist of dopamine and histamine receptors, exhibiting antipsychotic activity. This compound is intended for research use only and is not for patient administration. It is stable under recommended storage conditions.
- Antagonist of dopamine and histamine receptors
- Exhibits antipsychotic activity
- Purity of 99.91%
- Supplied as a solid, white to off-white in appearance
- Stable under recommended storage conditions
- Shipped at room temperature for short durations
- Recommended storage at 4°C, sealed, and away from moisture
- Soluble in water, ethanol, and DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.