Learn More
Medchemexpress LLC Chlorprothixene hydrochloride | 6469-93-8 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0274A500MG
Encompass_Preferred
Chlorprothixene hydrochloride functions as a dopamine and histamine receptor antagonist. It exhibits Ki values of 18 nM for hD1, 2.96 nM for hD2, 4.56 nM for hD3, 9 nM for hD5, and 3.75 nM for hH1 receptors, and also demonstrates antipsychotic activity.
  • Dopamine and histamine receptor antagonist.
  • Exhibits Ki values of 18 nM (hD1), 2.96 nM (hD2), 4.56 nM (hD3), 9 nM (hD5), and 3.75 nM (hH1).
  • Possesses antipsychotic activity.
  • Binds to 5-HT receptors with pKis of 8.3 (5-HT7), 8.5 (5-HT6), and 9.4 (5-HT2).
Catalog No. 50-004-09119