Learn More
Medchemexpress LLC Chlorprothixene hydrochloride | 6469-93-8 | 99.91% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0274A10MM/1ML
Encompass_Preferred
Chlorprothixene hydrochloride is an antagonist of dopamine and histamine receptors, exhibiting antipsychotic activity. It demonstrates specific Ki values for hD1, hD2, hD3, hD5, and hH1 receptors, and binds to 5-HT receptors with distinct pKis.
  • Antagonist of dopamine and histamine receptors
  • Exhibits antipsychotic activity
  • Binds to 5-HT receptors
  • Purity of 99.91%
Catalog No. 50-004-06449