Learn More
Medchemexpress LLC Chlorprothixene hydrochloride | 6469-93-8 | 99.91% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0274A100MG
Encompass_Preferred
Chlorprothixene hydrochloride is a dopamine and histamine receptor antagonist with Kis of 18 nM, 2.96 nM, 4.56 nM, 9 nM, and 3.75 nM for hD1, hD2, hD3, hD5, and hH1 receptors, respectively, exhibiting antipsychotic activity.
  • Dopamine and histamine receptor antagonist.
  • Antipsychotic activity.
  • High purity.
  • Soluble in water, ethanol, and DMSO.
  • Available in various quantities.
Catalog No. 50-004-07855