Learn More
Chem Service Inc Chlorpyrifos,2921-88-2,250G
SDP
Supplier:  Chem Service Inc N11459250MG
Encompass_Preferred
Molecular Formula C9H11Cl3O3PS, Purity 99.5%, Molecular Weight 350.59, Boiling Point 160 °C, Melting Point 41 - 42 °C. Chlorpyrifos is a crystalline organophosphate insecticide, acaracide and miticide.
Catalog No. NC0124229