Learn More
Medchemexpress LLC Chlorpyrifos-d10 | 285138-81-0 | 99.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0815S5MG
Encompass_Preferred
Chlorpyrifos-d10 is the deuterium labeled form of Chlorpyrifos, an organophosphate insecticide classified as a phosphorothionate. Its oxon metabolite inhibits acetylcholinesterase (AChE), which impacts neurological function in various organisms. The plasma enzyme paraoxonase 1 (PON1) hydrolyzes the Chlorpyrifos oxon (CPO) metabolite, and PON1 overexpression can mitigate susceptibility to neurotoxicity associated with CPO exposure. This compound is intended for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07626