Learn More
Chem Service Inc Chlorpyrifos Oxon,5598-15-2,100MG
SDP
Supplier:  Chem Service Inc MET11459B100MG
Encompass_Preferred
Purity 99%; Melting Point 41.3 °C; Synonyms: o-analog, Diethyl-3,5,6-trichloro-2-pyridinyl phosphate, Fospirate-ethyl, Dursbanoxon
Catalog No. NC9569833