Learn More
Medchemexpress LLC Chlorpyrifos-oxon | 5598-15-2 | 99.4% | 334.52 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13661010MM/1ML
Encompass_Preferred
Chlorpyrifos-oxon is an active metabolite of Chlorpyrifos, acting as a potent phosphorylating agent that inhibits Acetylcholinesterase (AChE). This compound can induce cross-linking between subunits of tubulin and disrupt microtubule function.
  • Potent phosphorylating agent
  • Inhibits Acetylcholinesterase (AChE)
  • Induces cross-linking between tubulin subunits
  • Disrupts microtubule function
  • Leads to protein aggregation in tubulin
  • Promotes isopeptide bond cross-linking of tubulin monomers
  • Inhibits neurite outgrowth
Catalog No. 50-003-51887