Learn More
Medchemexpress LLC Chlorpyrifos-oxon | 5598-15-2 | MFCD01705571 | 99.4% | C9H11Cl3NO4P | 10MG

Supplier: Medchemexpress LLC HY13661010MG
Chlorpyrifos-oxon is the oxon metabolite of the organophosphate insecticide chlorpyrifos. It is a phosphorylating agent and potent acetylcholinesterase inhibitor used in biochemical and toxicological research. The compound is characterized by formula C9H11Cl3NO4P, molecular weight 334.52, and is supplied at high purity for analytical and research applications.
- High purity (99.4%).
- Potent acetylcholinesterase inhibitor for mechanistic studies.
- Soluble in DMSO at 100 mg/mL.
- Physical form: solid below 43°C, liquid above 43°C.
- Long-term storage: stable at -20°C for up to 3 years in pure form.
- Formulation options available for in vivo use (DMSO/PEG300/Tween-80 or SBE-β-CD in saline).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.