Learn More
Medchemexpress LLC Chlorquinaldol | 72-80-0 | 98.7% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB13601G
Encompass_Preferred
Chlorquinaldol (Chloquinan) is an antibacterial agent that can be used for topical skin conditions and vaginal infections. It is also a β-catenin/TCF4 inhibitor, demonstrating anti-proliferation, anti-migration, and apoptosis-inducing activity in cancer cells.
  • Functions as an antibacterial agent
  • Used for topical skin conditions and vaginal infections
  • Acts as a β-catenin/TCF4 inhibitor
  • Demonstrates anti-proliferation activity in cancer cells
  • Shows anti-migration activity in cancer cells
  • Induces apoptosis in cancer cells
Catalog No. 50-004-12088