Learn More
Alkali Scientific Chlorsulfuron, 100 Milligrams
SDP
Supplier:  Alkali Scientific LOR001CP
Encompass_Preferred
Chlorsulfuron is a selective sulfonylurea herbicide used in laboratory settings for studying plant biochemistry and herbicide resistance. This high-purity 100mg pack of chlorsulfuron is used primarily in agricultural research, specifically in experiments related to herbicide resistance and the biochemical pathways of plant growth inhibition. Chlorsulfuron acts by inhibiting acetolactate synthase, an enzyme crucial for the synthesis of essential amino acids in plants. This mechanism of action makes it an effective tool in herbicide resistance studies and the development of weed management strategies in agriculture. It is also used in laboratory research focused on plant metabolic pathways.
Catalog No. 50-006-60027