Learn More
Selleck Chemical LLC Chlorzoxazone S4155-50mg
SDP
Supplier:  Selleck Chemical LLC S415550MG
Encompass_Preferred
Chlorzoxazone(Chlorzoxazon Paraflex) is a muscle-relaxing drug and a probe for human liver cytochrome P-450IIE1
Catalog No. 50-310-8540