Learn More
Medchemexpress LLC Chm-1 | 154554-41-3 | MFCD16627986 | 99.6% | 283.25 g·mol⁻¹ | C16H10FNO3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10325750MG
Encompass_Preferred
CHM-1 is a microtubule-destabilizing research compound that inhibits tubulin polymerization and exhibits potent antimitotic and antitumor activity in cellular studies, inducing G2-M phase arrest via activation of Cdc2 kinase.
  • Inhibits tubulin polymerization and disrupts microtubule dynamics.
  • Induces G2-M cell cycle arrest and promotes apoptosis in cancer cells.
  • Reported high purity (~99.6%) suitable for biochemical research applications.
  • Supplied in small milligram quantities for laboratory assays and screening.
  • Molecular weight 283.25 g·mol⁻¹; formula C16H10FNO3.
Catalog No. 50-002-46033