Learn More
Cygnus Technologies CHO DNA Concentrate, 1 mL
Supplier:  Cygnus Technologies D5521
Encompass
For use in spike and recovery experiments and as standards. Concentration of ~10 ug/ml.
Catalog No. NC0584556