Learn More
Eurofins Discoverx PathHunter CHO-K1 GPR120L (Long Isoform) β-Arrestin Cell Line, 2 vials. (93-0366C2)
Supplier:  Eurofins Discoverx 930366C2
Encompass
The PathHunter GPR120L cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of GPR120L GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR120L and EA-tagged β-Arrestin-2. Activation of GPR120L stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0362
Add to Cart
Request a Quote