Learn More
Cygnus Technologies CHO HCP ELISA 3G, Robotics Kit, 4 x 96 Well Plate
Supplier:  Cygnus Technologies F55014
Encompass
CHO HCP ELISA 3G, Robotics Kit is designed for automated workflows. This kit is a larger size of the F550-1 kit. It includes four Anti-CHO Coated Microtiter Strip plates and all reagents and standards in the volumes sufficient to perform CHO HCP ELISA on the Hamilton Microlab STAR™ Liquid Handling System. With a flexible deck layout, the Microlab STAR accommodates CHO HCP ELISA, 3G assay protocol. The complementary Microlab STAR’s VENUS software script to run the assay can be downloaded via the below provided link.
Catalog No. NC1911901