Learn More
Genscript Corporation CHO-K1/5-HT2A Stable Cell Line
Supplier:  Genscript Corporation M00251
Encompass
Recombinant CHO-K1 cells stably overexpress human 5-hydroxytryptamine receptor 2A (5-HT2A) on the surface and contain high levels of G protein G q and G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-59045
Add to Cart
Request a Quote