Learn More
Genscript Corporation CHO-K1/ADRA2A/G15 Stable Cell
Supplier:  Genscript Corporation M00281
Encompass
Recombinant CHO-K1 cells stably overexpress human adrenoceptor alpha 2A (ADRA2A) on the surface and contain high levels of G protein G i to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57736
Only null left
Add to Cart
Request a Quote