Learn More
Genscript Corporation CHO-K1/B7-H4 Stable Cell Line
Supplier:  Genscript Corporation M00537
Encompass
B7-H4 is a transmembrane protein that binds an unknown receptor on activated T cells resulting in inhibition of T-cell effector function via cell cycle arrest decreased proliferation and reduced IL-2 production B7-H4 is up-regulated on the surface of cancer cells and immunosuppressive tumor-associated macrophages (TAMs) in a variety of human cancers
Catalog No. 50-000-58736
Add to Cart
Request a Quote